missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat DLL1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
POA
Specifications
Antigen | DLL1 |
---|---|
Clone | 409005 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
DLL1 Monoclonal specifically detects DLL1 in Rat samples. It is validated for Western Blot.Specifications
DLL1 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Mouse | |
Rat | |
P97677 | |
28514 | |
Mouse myeloma cell line NS0-derived recombinant rat DLL1 Gln18-Trp537 Accession # P97677 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects rat DLL1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant rat (rr) Jagged-1, recombinant mouse (rm) Jagged-2, rmNotch-3, rrNotch-2, and rrNotch-1 is observed. |
409005 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Delta, delta (Drosophila)-like 1, Delta 1, Delta1, delta-like 1 (Drosophila), delta-like protein 1, DL1, Drosophila Delta homolog 1, H-Delta-1 | |
DLL1 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only