missing translation for 'onlineSavingsMsg'
Learn More

DL-TBOA, Tocris Bioscience™

Selective non-transportable inhibitor of EAATs

Brand:  Tocris (Bio-Techne) 1223/10

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13448643

  • £204.33 / 10mg

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

A competitive, non-transportable blocker of excitatory amino acid transporters (IC50 values are 70, 6, and 6 μM for EAAT1, EAAT2 and EAAT3 respectively). Also inhibits EAAT4 and EAAT5 (Ki values are 4.4 and 3.2 μM respectively). Displays high selectivity for EAATs over ionotropic and metabotropic glutamate receptors. Also available as part of the Excitatory Amino Acid Transporter Inhibitor Tocriset™ .
Specifications

Chemical Identifiers

205309-81-5
239.227
dl-tboa, chembl79140, dl-threo-beta-benzyloxyaspartate, 3s-3-benzyloxy-l-aspartic acid, 2s,3s-2-amino-3-benzyloxy succinic acid, tboa, 2s,3s-2-amino-3-benzyloxy-succinic acid, tocris-1223, d07gub, l-threo-benzyloxy aspartate
(2S,3S)-2-amino-3-phenylmethoxybutanedioic acid
C11H13NO5
BYOBCYXURWDEDS-IUCAKERBSA-N
5311218
C1=CC=C(C=C1)COC(C(C(=O)O)N)C(=O)O

Specifications

DL-TBOA
C11H13NO5
BYOBCYXURWDEDS-IUCAKERBSA-N
(2S,3S)-2-amino-3-phenylmethoxybutanedioic acid
5311218
>98%
205309-81-5
dl-tboa, chembl79140, dl-threo-beta-benzyloxyaspartate, 3s-3-benzyloxy-l-aspartic acid, 2s,3s-2-amino-3-benzyloxy succinic acid, tboa, 2s,3s-2-amino-3-benzyloxy-succinic acid, tocris-1223, d07gub, l-threo-benzyloxy aspartate
C1=CC=C(C=C1)COC(C(C(=O)O)N)C(=O)O
239.227
239.23
10mg
Videos
Documents