missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals DL-Selenomethionine, 99+%
Click to view available options
Quantity:
500 mg
1 g
2.5 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 265.0°C |
Color | Colorless |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% min. (HPLC) |
Odor | Faint Smelling |
Packaging | Glass bottle |
Linear Formula | CH3SeCH2CH2CH(NH2)COOH |
Quantity | 2.5 g |
Merck Index | 15, 8578 |
Solubility Information | Solubility in water: soluble. Other solubilities: soluble in methanol, ethanol, insoluble in benzene, diethyl ether |
Show More |
Safety and Handling
Product Identifier
- DL-Selenomethionine
- Danger
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Specific target organ toxicity after repeated exposure Category 2
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H373-May cause damage to organs.
- H331-Toxic if inhaled.
- H301-Toxic if swallowed.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'