missing translation for 'onlineSavingsMsg'
Learn More

DL-Propargylglycine, 98%, Thermo Scientific Chemicals

Catalog Number 10472560
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
64165-64-6
C5H7NO2
113.12
MFCD00056728
DGYHPLMPMRKMPD-UHFFFAOYNA-N
dl-propargylglycine, 2-amino-4-pentynoic acid, h-dl-pra-oh, 2-azanylpent-4-ynoic acid, dl-2-propynylglycine, 2-propynylglycine, l-2-amino-4-pentynoic acid, d,l-propargylglycine, propargyl glycine, +--2-amino-4-pentynoic acid
95575
2-aminopent-4-ynoic acid
NC(CC#C)C(O)=O
This item is not returnable. View return policy
64165-64-6
C5H7NO2
113.12
MFCD00056728
DGYHPLMPMRKMPD-UHFFFAOYNA-N
dl-propargylglycine, 2-amino-4-pentynoic acid, h-dl-pra-oh, 2-azanylpent-4-ynoic acid, dl-2-propynylglycine, 2-propynylglycine, l-2-amino-4-pentynoic acid, d,l-propargylglycine, propargyl glycine, +--2-amino-4-pentynoic acid
95575
2-aminopent-4-ynoic acid
NC(CC#C)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 235.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 113.12
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material DL-Propargylglycine
compliance-icons
Product Identifier
  • DL-Propargylglycine, 98%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'