missing translation for 'onlineSavingsMsg'
Learn More

DL-Pantolactone, 97%, ACROS Organics™

Brand:  Acros Organics 427591000

Code : 29

Additional Details : CAS Number : 79-50-5 Weight : 0.15000kg


Quantity 100g
Packaging Glass bottle

 View more versions of this product

Product Code. 10507644

  • £96.40 / 100g


Estimated Shipment
19-06-2018
Add to basket


Specifications

Chemical Identifiers

DL-Pantolactone
C6H10O3
MFCD00064333
97%
130.14

Specifications

DL-Pantolactone
97%
C6H10O3
MFCD00064333
Solubility in water: soluble.
246.0°C
122°C
79-50-5
97%
130.14
Glass bottle
74.0° to 78.0°C
100g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • DL-Pantolactone

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS