missing translation for 'onlineSavingsMsg'
Learn More

DL-o-Tyrosine, 98%, Thermo Scientific™

Catalog Number p-7059408
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
2370-61-8
C9H11NO3
181.19
MFCD00021725
WRFPVMFCRNYQNR-UHFFFAOYNA-N
dl-o-tyrosine, 2-amino-3-2-hydroxyphenyl propanoic acid, o-tyrosine, 2-hydroxyphenylalanine, 2-hydroxy-dl-phenylalanine, ortho-tyrosine, 2-tyrosine, o-tyrosine, dl, dl-3-o-hydroxyphenyl alanine, 2-amino-3-2-hydroxyphenyl propionic acid
91482
2-amino-3-(2-hydroxyphenyl)propanoic acid
NC(CC1=CC=CC=C1O)C(O)=O
This item is not returnable. View return policy
2370-61-8
C9H11NO3
181.19
MFCD00021725
WRFPVMFCRNYQNR-UHFFFAOYNA-N
dl-o-tyrosine, 2-amino-3-2-hydroxyphenyl propanoic acid, o-tyrosine, 2-hydroxyphenylalanine, 2-hydroxy-dl-phenylalanine, ortho-tyrosine, 2-tyrosine, o-tyrosine, dl, dl-3-o-hydroxyphenyl alanine, 2-amino-3-2-hydroxyphenyl propionic acid
91482
2-amino-3-(2-hydroxyphenyl)propanoic acid
NC(CC1=CC=CC=C1O)C(O)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point ∼256°C (decomposition)
Beilstein 2805789
Solubility Information Soluble in water,and DMSO (Heated).
Formula Weight 181.19
Percent Purity 98%
Quantity 1 g
Physical Form Powder
Chemical Name or Material DL-o-Tyrosine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'