missing translation for 'onlineSavingsMsg'
Learn More

DL-α-(Methylaminomethyl)benzyl alcohol, 99%, ACROS Organics™

Brand:  Acros Organics 198230010

Code : 29

Additional Details : CAS Number : 68579-60-2 Weight : 0.05100kg Transport : UN number : 3259 Chem class : 1b Pack group : III


Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10338720

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

DL-α-(Methylaminomethyl)benzyl alcohol
6589-55-5
99%
C6H5CH(CH2NHCH3)OH
MFCD00004506
99%
99%
C9H13NO
151.21
2-Methylamino-1-phenylethanol

Specifications

DL-α-(Methylaminomethyl)benzyl alcohol
74.0° to 76.0°C
99%
Authentic
Fine Crystalline Needles
99%
C6H5CH(CH2NHCH3)OH
MFCD00004506
2-Methylamino-1-phenylethanol
White
1g
6589-55-5
Glass bottle
99%
C9H13NO
151.21
10, 4485
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • DL-a-(Methylaminomethyl)benzyl alcohol

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.

SDS