missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ DL-Menthyl borate, 97%

Brand:  Alfa Aesar™ 043709.06

Code : NEW

Additional Details : CAS Number : 62697-74-9 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 11390308

  • £67.30 / 5g


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Specifications

Chemical Identifiers

DL-Menthyl borate
C30H57BO3
MFCD00041735
62697-74-9
476.58

Specifications

DL-Menthyl borate
5g
Crystalline
C30H57BO3
MFCD00041735
White
62697-74-9
97%
476.58
Soluble in chloroform, ether
Videos
SDS