missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ DL-Menthyl borate, 97%

Brand:  Alfa Aesar™ 043709.03

Code : NEW

Additional Details : CAS Number : 62697-74-9 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 11380308

  • £19.00 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

DL-Menthyl borate
C30H57BO3
MFCD00041735
62697-74-9
476.58

Specifications

DL-Menthyl borate
1g
Crystalline
C30H57BO3
MFCD00041735
White
62697-74-9
97%
476.58
Soluble in chloroform, ether
Videos
SDS