missing translation for 'onlineSavingsMsg'
Learn More

DL-Menthol, 99%, Thermo Scientific Chemicals

Catalog Number p-155566
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
89-78-1
C10H20O
156.27
MFCD00001484
NOOLISFMXDJSKH-IVZWLZJFSA-N
menthol, dl-menthol, 2-isopropyl-5-methylcyclohexanol, menthyl alcohol, cyclohexanol, 5-methyl-2-1-methylethyl, p-menthan-3-ol, racementhol, caswell no. 540, 3-p-menthol, +-neo-menthol
6566020
(1R,2R,5S)-5-methyl-2-propan-2-ylcyclohexan-1-ol
CC1CCC(C(C1)O)C(C)C
This item is not returnable. View return policy
89-78-1
C10H20O
156.27
MFCD00001484
NOOLISFMXDJSKH-IVZWLZJFSA-N
menthol, dl-menthol, 2-isopropyl-5-methylcyclohexanol, menthyl alcohol, cyclohexanol, 5-methyl-2-1-methylethyl, p-menthan-3-ol, racementhol, caswell no. 540, 3-p-menthol, +-neo-menthol
6566020
(1R,2R,5S)-5-methyl-2-propan-2-ylcyclohexan-1-ol
CC1CCC(C(C1)O)C(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 32.0°C to 36.0°C
Color Colorless to White
Density 0.8900g/mL
Boiling Point 216.0°C
Flash Point 92°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 5 mL
Merck Index 15, 5905
Specific Gravity 0.89
Specific Rotation Condition −2.00 (20.00°C c=10,EtOH)
Specific Rotation − 2.00
Solubility Information Solubility in water: insoluble. Other solubilities: freely soluble in glacial acetic acid,soluble in alcohol,chloroform and ether
Formula Weight 156.27
Percent Purity 99%
Physical Form Crystalline Solid
Chemical Name or Material DL-Menthol, 99%
Show More Show Less
compliance-icons
Product Identifier
  • DL-Menthol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'