missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals DL-Lysine monohydrochloride, 99%
Click to view available options
Quantity:
25 g
100 g
Description
DL-Lysine monohydrochloride acts as an intermediate in the preparation of 3-amino-azepan-2-one. It also serves as an essential amino acid. It is used as animal feed. In addition to this, it is used in the construction of polylysine polymers, which finds application in lysine-based polypeptide gels and films.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
DL-Lysine monohydrochloride acts as an intermediate in the preparation of 3-amino-azepan-2-one. It also serves as an essential amino acid. It is used as animal feed. In addition to this, it is used in the construction of polylysine polymers, which finds application in lysine-based polypeptide gels and films.
Solubility
Soluble in water. Insoluble in ethanol, ethyl ether, acetone, benzene and common neutral solvents.
Notes
Incompatible with strong oxidizing agents.
DL-Lysine monohydrochloride acts as an intermediate in the preparation of 3-amino-azepan-2-one. It also serves as an essential amino acid. It is used as animal feed. In addition to this, it is used in the construction of polylysine polymers, which finds application in lysine-based polypeptide gels and films.
Solubility
Soluble in water. Insoluble in ethanol, ethyl ether, acetone, benzene and common neutral solvents.
Notes
Incompatible with strong oxidizing agents.
Specifications
Melting Point | 267°C (decomposition) |
Beilstein | 4711993 |
Solubility Information | Soluble in water. Insoluble in ethanol,ethyl ether,acetone,benzene and common neutral solvents. |
Formula Weight | 182.65 |
Percent Purity | 99% |
Quantity | 25 g |
Chemical Name or Material | DL-Lysine monohydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'