missing translation for 'onlineSavingsMsg'
Learn More

DL-Isocitric acid, trisodium salt hydrate, 95%, Thermo Scientific Chemicals

Catalog Number p-3736716
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
1637-73-6
C6H5Na3O7
258.07
MFCD00149284
HWMVXEKEEAIYGB-UHFFFAOYNA-K
dl-isocitric acid, isocitric acid trisodium salt, sodium isocitrate, dl-isocitric acid trisodium salt hydrate, dl-isocitric acid trisodium salt, trisodium 1-hydroxypropane-1,2,3-tricarboxylate, trisodium isocitrate, tripotassium isocitrate, isocitric acid, dl, dl-isocitric acid, trisodium salt hydrate
168942
trisodium;1-hydroxypropane-1,2,3-tricarboxylate
[Na+].[Na+].[Na+].OC(C(CC([O-])=O)C([O-])=O)C([O-])=O
1637-73-6
C6H5Na3O7
258.07
MFCD00149284
HWMVXEKEEAIYGB-UHFFFAOYNA-K
dl-isocitric acid, isocitric acid trisodium salt, sodium isocitrate, dl-isocitric acid trisodium salt hydrate, dl-isocitric acid trisodium salt, trisodium 1-hydroxypropane-1,2,3-tricarboxylate, trisodium isocitrate, tripotassium isocitrate, isocitric acid, dl, dl-isocitric acid, trisodium salt hydrate
168942
trisodium;1-hydroxypropane-1,2,3-tricarboxylate
[Na+].[Na+].[Na+].OC(C(CC([O-])=O)C([O-])=O)C([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula NaO2CCH2CH(CO2Na)CH(OH)CO2Na·xH2O
Quantity 1 g
Beilstein 03, 555
Solubility Information Solubility in water: soluble in water
Formula Weight 258.07
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material DL-Isocitric acid, trisodium salt hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'