missing translation for 'onlineSavingsMsg'
Learn More

DL-Isoborneol, 93%, tech., Thermo Scientific Chemicals

Catalog Number p-150229
Click to view available options
Quantity:
25 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
124-76-5
C10H18O
154.25
MFCD00074821
DTGKSKDOIYIVQL-SZBHIRRCSA-N
dl-isoborneol
126961757
(1R,3R)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-ol
CC1(C2CCC1(C(C2)O)C)C
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
124-76-5
C10H18O
154.25
MFCD00074821
DTGKSKDOIYIVQL-SZBHIRRCSA-N
dl-isoborneol
126961757
(1R,3R)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-ol
CC1(C2CCC1(C(C2)O)C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 208.0°C to 214.0°C
Color Yellow
Flash Point 74°C
Infrared Spectrum Authentic
Assay Percent Range 91% min. (GC)
Packaging Plastic bottle
Quantity 25 g
Beilstein 06, II, 80
Merck Index 15, 5173
Solubility Information Solubility in water: insoluble in water. Other solubilities: readily soluble in alcohol,chloroform and ether
Formula Weight 154.25
Percent Purity 93%
Grade Technical
Physical Form Powder
Chemical Name or Material DL-Isoborneol, tech., 93%
Show More Show Less
compliance-icons
Product Identifier
  • DL-Isoborneol
Signal Word
  • Warning
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'