missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Glutamic acid monohydrate, 98+%

Catalog Number p-138153
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
19285-83-7
C5H9NO4·H2O
165.15
MFCD00150703
OZDAOHVKBFBBMZ-UHFFFAOYSA-N
dl-glutamic acid monohydrate, 2-aminopentanedioic acid hydrate, dl-glutamic acid hydrate, h-dl-glu-oh.h2o, glutaminsaeure hydrate, +/--2-aminoglutaric acid, dl-2-aminopentanedioic acid, 2-azanylpentanedioic acid hydrate, 2-aminopentanedioic acid, oxamethane, hydrate, a-aminoglutarate
16219385
2-aminopentanedioic acid;hydrate
C(CC(=O)O)C(C(=O)O)N.O
This item is not returnable. View return policy
19285-83-7
C5H9NO4·H2O
165.15
MFCD00150703
OZDAOHVKBFBBMZ-UHFFFAOYSA-N
dl-glutamic acid monohydrate, 2-aminopentanedioic acid hydrate, dl-glutamic acid hydrate, h-dl-glu-oh.h2o, glutaminsaeure hydrate, +/--2-aminoglutaric acid, dl-2-aminopentanedioic acid, 2-azanylpentanedioic acid hydrate, 2-aminopentanedioic acid, oxamethane, hydrate, a-aminoglutarate
16219385
2-aminopentanedioic acid;hydrate
C(CC(=O)O)C(C(=O)O)N.O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 194.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Plastic bottle
Linear Formula HO2CCH2CH2CH(NH2)CO2H·H2O
Quantity 25 g
Solubility Information Solubility in water: 20.54g/L (25°C). Other solubilities: sparingly soluble in alcohol and ether
Formula Weight 165.15
Percent Purity 98+%
Physical Form Powder
Chemical Name or Material DL-Glutamic acid monohydrate, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'