missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Ethionine, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
2.5 g
10 g
50 g
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 265°C to 269°C |
Color | White |
Assay Percent Range | 98% |
Infrared Spectrum | Authentic |
Linear Formula | C2H5SCH2CH2CH(NH2)CO2H |
Merck Index | 15, 3792 |
Packaging | Glass Bottle |
Formula Weight | 163.23 |
Percent Purity | ≥97.5% |
Quantity | 10 g |
Show More |
Safety and Handling
Product Identifier
- DL-Ethionine
- Warning
- Carcinogenicity Category 2
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 2
- H351-Suspected of causing cancer.
- H361f-Suspected of damaging fertility.
- H341-Suspected of causing genetic defects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'