missing translation for 'onlineSavingsMsg'
Learn More

DL-Cysteine hydrochloride hydrate, 99%, ACROS Organics™

Brand:  Acros Organics 227080100

Code : 29

Additional Details : CAS Number : 10318-18-0 Weight : 0.06000kg


Quantity 10g

 View more versions of this product

Product Code. 10236760

  • £101.90 / 10g


EU Stock 1
Add to basket


Specifications

Chemical Identifiers

DL-Cysteine hydrochloride hydrate
96998-61-7
99%
HCl·xH2O
99%
98.5 to 101.5% (on dried substance)
HCl·xH2O
MFCD00151030

Specifications

DL-Cysteine hydrochloride hydrate
96998-61-7
10ppm max.
98.5 to 101.5% (on dried substance)
Authentic
HCl·xH2O
8.5% to 12% (vacuum, P2O5, room temp., 20 hrs)
Glass bottle
98% min. (c=10, H2O, 430nm) 1cm cell
White
99%
1ppm max.
Crystals or Crystalline Powder
99%
10ppm max.
HCl·xH2O
MFCD00151030
280ppm max.
0.1% max.
10g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • DL-Cysteine hydrochloride hydrate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/A§.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS