missing translation for 'onlineSavingsMsg'
Learn More

DL-Citrulline, 97%, Thermo Scientific™

Catalog Number p-7081417
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
627-77-0
C6H13N3O3
175.188
MFCD00007955
RHGKLRLOHDJJDR-UHFFFAOYSA-N
dl-citrulline, citrullin, 2-amino-5-ureidopentanoic acid, dl-2-amino-5-ureidovaleric acid, citrulina, citrulline, dl, ornithine, n5-carbamoyl-, dl, dl-ornithine, n 5-aminocarbonyl, n 5-carbamoyl-dl-ornithine, n 5-aminocarbonyl ornithine
833
CHEBI:18211
2-amino-5-(carbamoylamino)pentanoic acid
C(CC(C(=O)O)N)CNC(=O)N
This item is not returnable. View return policy
627-77-0
C6H13N3O3
175.188
MFCD00007955
RHGKLRLOHDJJDR-UHFFFAOYSA-N
dl-citrulline, citrullin, 2-amino-5-ureidopentanoic acid, dl-2-amino-5-ureidovaleric acid, citrulina, citrulline, dl, ornithine, n5-carbamoyl-, dl, dl-ornithine, n 5-aminocarbonyl, n 5-carbamoyl-dl-ornithine, n 5-aminocarbonyl ornithine
833
CHEBI:18211
2-amino-5-(carbamoylamino)pentanoic acid
C(CC(C(=O)O)N)CNC(=O)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 5 g
Formula Weight 175.19
Percent Purity 97%
Chemical Name or Material DL-Citrulline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'