missing translation for 'onlineSavingsMsg'
Learn More

DL-Buthionine (S,R)-sulfoximine, 99%, Thermo Scientific Chemicals

Catalog Number 10480151
Click to view available options
Quantity:
500 mg
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
5072-26-4
C8H18N2O3S
222.30
MFCD00070309
KJQFBVYMGADDTQ-UHFFFAOYNA-N
buthionine sulfoximine, dl-buthionine-s,r-sulfoximine, buthionine sulfoxamine, buthione sulfoximine, buthionine-s,r-sulfoximine, 2-amino-4-s-butylsulfonimidoyl butanoic acid, butionine sulfoximine, buthionine sulphoximine, 2-amino-4-butylsulfonimidoyl butanoic acid, butanoic acid, 2-amino-4-s-butylsulfonimidoyl
21157
CHEBI:28714
2-amino-4-(butylsulfonimidoyl)butanoic acid
CCCCS(=N)(=O)CCC(N)C(O)=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
5072-26-4
C8H18N2O3S
222.30
MFCD00070309
KJQFBVYMGADDTQ-UHFFFAOYNA-N
buthionine sulfoximine, dl-buthionine-s,r-sulfoximine, buthionine sulfoxamine, buthione sulfoximine, buthionine-s,r-sulfoximine, 2-amino-4-s-butylsulfonimidoyl butanoic acid, butionine sulfoximine, buthionine sulphoximine, 2-amino-4-butylsulfonimidoyl butanoic acid, butanoic acid, 2-amino-4-s-butylsulfonimidoyl
21157
CHEBI:28714
2-amino-4-(butylsulfonimidoyl)butanoic acid
CCCCS(=N)(=O)CCC(N)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 215°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3(CH2)3S(O)(=NH)CH2CH2CH(NH2)CO2H
Quantity 500 mg
Merck Index 15, 1528
Solubility Information Solubility in water: soluble
Formula Weight 222.31
Percent Purity 99%
Physical Form Fluffy Powder
Chemical Name or Material DL-Buthionine (S, R)-sulfoximine, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'