missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ DL-Arginine monohydrochloride monohydrate, 98+%, Thermo Scientific™

Catalog Number p-7080981
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
332360-01-7
C6H17ClN4O3
228.677
MFCD00151031
NUFCANQNMWVENN-UHFFFAOYSA-N
dl-arginine hydrochloride monohydrate, 2-amino-5-guanidinopentanoic acid hydrochloride hydrate, 2-amino-5-diaminomethylideneamino pentanoic acid hydrate hydrochloride, h-dl-arg-oh.hcl, arginine hydrate hydrochloride, pubchem12368, acmc-1ajcb, arginine, chloride, hydrate, dl-arginine monohydrochloride 1-hydrate, dl-arginine monohydrochloride monohydrate
2734011
2-amino-5-(diaminomethylideneamino)pentanoic acid;hydrate;hydrochloride
C(CC(C(=O)O)N)CN=C(N)N.O.Cl
This item is not returnable. View return policy
332360-01-7
C6H17ClN4O3
228.677
MFCD00151031
NUFCANQNMWVENN-UHFFFAOYSA-N
dl-arginine hydrochloride monohydrate, 2-amino-5-guanidinopentanoic acid hydrochloride hydrate, 2-amino-5-diaminomethylideneamino pentanoic acid hydrate hydrochloride, h-dl-arg-oh.hcl, arginine hydrate hydrochloride, pubchem12368, acmc-1ajcb, arginine, chloride, hydrate, dl-arginine monohydrochloride 1-hydrate, dl-arginine monohydrochloride monohydrate
2734011
2-amino-5-(diaminomethylideneamino)pentanoic acid;hydrate;hydrochloride
C(CC(C(=O)O)N)CN=C(N)N.O.Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Beilstein 4158960
Formula Weight 228.69 (210.67 Anhydrous)
Percent Purity ≥98%
Quantity 5 g
Chemical Name or Material DL-Arginine monohydrochloride monohydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'