DL-Arginine hydrochloride, 98%, anhydrous, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 446020050

UNSPSC: 12161500

Code: 29

Additional Details:
Additional Details: CAS Number: 32042-43-6 Weight: 0.05500kg


Quantity 5g

Product Code. 11905541

Quantity Price
1 £ 41.4 / 5g
Estimated Shipment date
from Supplier 10-04-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material DL-Arginine hydrochloride
Assay 97.5 to 102.5%
Assay Percent Range 98%
Molecular Formula C6H14N4O2·ClH
Linear Formula H2NC(=NH)NH(CH2)3CH(NH2)CO2H·HCl
Formula Weight 210.67
Synonym DL-2-Amino-5-guanidinopentanoic acid hydrochloride monohydrate

Specification

Physical Form Powder
Formula Weight 210.67
Quantity 5g
Color White to off-white
Assay 97.5 to 102.5%
Linear Formula H2NC(=NH)NH(CH2)3CH(NH2)CO2H·HCl
CAS 32042-43-6
Infrared Spectrum Authentic
Loss on Drying 3% max.
Assay Percent Range 98%
Molecular Formula C6H14N4O2·ClH
Synonym DL-2-Amino-5-guanidinopentanoic acid hydrochloride monohydrate
Chemical Name or Material DL-Arginine hydrochloride
Documentation

Technical documents