Learn More
DL-AP5, Tocris Bioscience™
Potent, selective NMDA antagonist
Brand:  Tocris (Bio-Techne) 0105/10
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13487423
  • £79.65 / 10mg
Estimated Shipment
04-10-2018
Add to basket
Description
Description
Potent NMDA antagonist. Separate isomers D-AP5 (Cat. No. 0106), L-AP5 (Cat. No. 0107) and sodium salt DL-AP5 Sodium salt (Cat. No. 3693) also available.
Specifications
Chemical Identifiers
76326-31-3
197.127
DL-2-Amino-5-phosphonopentanoic acid
CHEBI:138644
C(CC(C(=O)O)N)CP(=O)(O)O
C5H12NO5P
VOROEQBFPPIACJ-UHFFFAOYSA-N
1216
2-amino-5-phosphonopentanoic acid
Specifications
DL-AP5
>98%
C5H12NO5P
VOROEQBFPPIACJ-UHFFFAOYSA-N
2-amino-5-phosphonopentanoic acid
1216
197.13
76326-31-3
10mg
DL-2-Amino-5-phosphonopentanoic acid
C(CC(C(=O)O)N)CP(=O)(O)O
197.127
CHEBI:138644
Videos