Learn More
Tocris Bioscience™ DL-AP5 Sodium salt
Potent and selective NMDA antagonist; sodium salt of DL-AP5
Brand:  Tocris Bioscience™ 369350
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13443434
  • £278.53 / 50mg
Estimated Shipment
27-06-2019
Description
Description
Sodium salt of DL-AP5 (Cat. No. 0105). Potent NMDA antagonist.
Specifications
Specifications
DL-AP5 Sodium salt
>98
50mg
DL-AP5 Sodium salt
219.11