missing translation for 'onlineSavingsMsg'
Learn More

DL-Aminoglutethimide, 99%, Thermo Scientific Chemicals

Catalog Number p-7058214
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
125-84-8
C13H16N2O2
232.283
MFCD00010122
ROBVIMPUHSLWNV-UHFFFAOYSA-N
aminoglutethimide, dl-aminoglutethimide, cytadren, orimeten, p-aminoglutethimide, elipten, 3-4-aminophenyl-3-ethylpiperidine-2,6-dione, aminoglutetimida, 3-4-aminophenyl-3-ethyl-2,6-piperidinedione, aminoglutethimidum
2145
CHEBI:2654
3-(4-aminophenyl)-3-ethylpiperidine-2,6-dione
CCC1(CCC(=O)NC1=O)C2=CC=C(C=C2)N
This item is not returnable. View return policy
125-84-8
C13H16N2O2
232.283
MFCD00010122
ROBVIMPUHSLWNV-UHFFFAOYSA-N
aminoglutethimide, dl-aminoglutethimide, cytadren, orimeten, p-aminoglutethimide, elipten, 3-4-aminophenyl-3-ethylpiperidine-2,6-dione, aminoglutetimida, 3-4-aminophenyl-3-ethyl-2,6-piperidinedione, aminoglutethimidum
2145
CHEBI:2654
3-(4-aminophenyl)-3-ethylpiperidine-2,6-dione
CCC1(CCC(=O)NC1=O)C2=CC=C(C=C2)N

Aminoglutethimide is used to decrease the production of sex hormones and suppress the growth of tumors that need sex hormones to grow. It blocks the production of steroids derived from cholesterol and is clinically used in the treatment of Cushing's syndrome and metastatic breast cancer. It is also a drug of abuse by body builders.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water (2 mg/ml at 20°C), methanol (50 mg/ml), ethanol (7 mg/ml at 25°C), DMSO (20 mg/ml at 25°C), and chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 152°C to 154°C
Color White
Quantity 1 g
Merck Index 14,440
Solubility Information Soluble in water (2mg/ml at 20°C),methanol (50mg/ml),ethanol (7mg/ml at 25°C),DMSO (20mg/ml at 25°C),and chloroform.
Formula Weight 232.28
Percent Purity 99%
Physical Form Powder
Chemical Name or Material DL-Aminoglutethimide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'