missing translation for 'onlineSavingsMsg'
Learn More

DL-Alanine methyl ester hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog Number p-3776561
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
13515-97-4
C4H10ClNO2
139.58
MFCD00035523
IYUKFAFDFHZKPI-UHFFFAOYNA-N
methyl 2-aminopropanoate hydrochloride, dl-alanine methyl ester hydrochloride, h-dl-ala-ome.hcl, h-dl-ala-ome hydrochloride, dl-alanine methyl ester hcl, methyl l-alaninate hcl, methyl dl-alaninate hcl, methyl dl-alaninate hydrochloride, h-dl-ala-ome hcl, methyl dl-2-aminopropanoate hydrochloride
2756298
Cl.COC(=O)C(C)N
This item is not returnable. View return policy
13515-97-4
C4H10ClNO2
139.58
MFCD00035523
IYUKFAFDFHZKPI-UHFFFAOYNA-N
methyl 2-aminopropanoate hydrochloride, dl-alanine methyl ester hydrochloride, h-dl-ala-ome.hcl, h-dl-ala-ome hydrochloride, dl-alanine methyl ester hcl, methyl l-alaninate hcl, methyl dl-alaninate hcl, methyl dl-alaninate hydrochloride, h-dl-ala-ome hcl, methyl dl-2-aminopropanoate hydrochloride
2756298
Cl.COC(=O)C(C)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Assay Percent Range 98+%
Infrared Spectrum Authentic
Linear Formula CH3CH(NH2)COOCH3·HCl
Packaging Glass Bottle
IUPAC Name methyl 2-aminopropanoate;hydrochloride
Formula Weight 139.58
Percent Purity ≥98.0%
Quantity 1 g
Physical Form Powder
Chemical Name or Material DL-Alanine methyl ester hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • DL-Alanine methyl ester hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'