missing translation for 'onlineSavingsMsg'
Learn More

DL-6-Hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrate, 96%, Thermo Scientific™

Catalog Number 10721355
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
76824-99-2
C10H11NO3
193.20
MFCD03092912
CRAGDYRHPWTZJL-UHFFFAOYNA-N
6-hydroxy-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid hydrate, 6-hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrate, akos 68, d,l-6-hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrate, 6-hydroxy-1,2,3,4-tetrahydro-isoquinoline-3-carboxylic acid h2o, 6-oxidanyl-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrate, d,l-6-hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
18525693
OC(=O)C1CC2=C(CN1)C=CC(O)=C2
This item is not returnable. View return policy
76824-99-2
C10H11NO3
193.20
MFCD03092912
CRAGDYRHPWTZJL-UHFFFAOYNA-N
6-hydroxy-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid hydrate, 6-hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrate, akos 68, d,l-6-hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrate, 6-hydroxy-1,2,3,4-tetrahydro-isoquinoline-3-carboxylic acid h2o, 6-oxidanyl-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrate, d,l-6-hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
18525693
OC(=O)C1CC2=C(CN1)C=CC(O)=C2
CAS Min % 95.0
CAS Max % 100.0
Melting Point 270°C to 276°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Quantity 1 g
IUPAC Name 6-hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
Formula Weight 211.22
Percent Purity ≥95%
Physical Form Crystalline Powder
Chemical Name or Material DL-6-Hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'