DL-5-Methoxytryptophan, 95%, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 151761000

100MG DL-5-Methoxytryptophan, 95%

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 28052-84-8 Weight: 0.05010kg


Packaging Glass bottle
Quantity 100mg

Product Code. 10206010

Quantity Price
1 £ 30.8 / 100mg
Estimated Shipment date
from Supplier 28-02-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material DL-5-Methoxytryptophan
CAS 28052-84-8
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C12H14N2O3
Formula Weight 234.25
MDL Number MFCD00005650

Specification

Assay ≥94%
Physical Form Powder
Formula Weight 234.25
Quantity 100mg
Color Off-white
Appearance of Solution (1% soln. in warm water) clear, colorless
Infrared Spectrum Authentic
CAS 28052-84-8
Assay Percent Range 95%
MDL Number MFCD00005650
Packaging Glass bottle
Water 3% max. (K.F.)
Molecular Formula C12H14N2O3
Chemical Name or Material DL-5-Methoxytryptophan
Documentation

Technical documents