missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-5-Hydroxytryptophan, 99%

Catalog Number p-7082009
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
56-69-9
C11H12N2O3
220.23
MFCD00005651
LDCYZAJDBXYCGN-UHFFFAOYSA-N
5-hydroxy-dl-tryptophan, 5-hydroxytryptophan, dl-5-hydroxytryptophan, 2-amino-3-5-hydroxy-1h-indol-3-yl propanoic acid, +--5-hydroxytryptophan, dl-hydroxytryptophan, dl-5-htp, 5-hydroxytryptophan dl-form, h-dl-trp 5-oh-oh, dl-pretonine
144
CHEBI:28171
2-amino-3-(5-hydroxy-1H-indol-3-yl)propanoic acid
NC(CC1=CNC2=CC=C(O)C=C12)C(O)=O
This item is not returnable. View return policy
56-69-9
C11H12N2O3
220.23
MFCD00005651
LDCYZAJDBXYCGN-UHFFFAOYSA-N
5-hydroxy-dl-tryptophan, 5-hydroxytryptophan, dl-5-hydroxytryptophan, 2-amino-3-5-hydroxy-1h-indol-3-yl propanoic acid, +--5-hydroxytryptophan, dl-hydroxytryptophan, dl-5-htp, 5-hydroxytryptophan dl-form, h-dl-trp 5-oh-oh, dl-pretonine
144
CHEBI:28171
2-amino-3-(5-hydroxy-1H-indol-3-yl)propanoic acid
NC(CC1=CNC2=CC=C(O)C=C12)C(O)=O

It is an immediate precursor that is used to assess central serotonergic function. It was used for the treatment of insomnia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an immediate precursor that is used to assess central serotonergic function. It was used for the treatment of insomnia.

Solubility
Soluble in water (4 mg/ml at 25°C), methanol.

Notes
Store at 4°C. Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Melting Point ∼298°C (decomposition)
Beilstein 88199
Merck Index 14,4847
Solubility Information Soluble in water (4mg/ml at 25°C),methanol.
Formula Weight 220.23
Percent Purity 99%
Quantity 1 g
Chemical Name or Material DL-5-Hydroxytryptophan

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'