missing translation for 'onlineSavingsMsg'
Learn More

DL-4-Fluoro-alpha-methylbenzylamine, 97%, Thermo Scientific™

Catalog Number p-134784
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
403-40-7
C8H10FN
139.17
MFCD00041323
QGCLEUGNYRXBMZ-UHFFFAOYSA-N
1-4-fluorophenyl ethanamine, 1-4-fluorophenyl ethylamine, 4-fluoro-alpha-methylbenzylamine, 1-4'-fluorophenyl ethanamine, dl-4-fluoro-alpha-methylbenzylamine, 4-fluoro-a-methylbenzylamine, 1-p-fluorophenyl-ethylamine, 1-4-fluorophenyl ethan-1-amine, p-fluoro-a-methylbenzylamine, rs-1-4-fluorophenyl ethylamine
123063
1-(4-fluorophenyl)ethanamine
CC(C1=CC=C(C=C1)F)N
This item is not returnable. View return policy
403-40-7
C8H10FN
139.17
MFCD00041323
QGCLEUGNYRXBMZ-UHFFFAOYSA-N
1-4-fluorophenyl ethanamine, 1-4-fluorophenyl ethylamine, 4-fluoro-alpha-methylbenzylamine, 1-4'-fluorophenyl ethanamine, dl-4-fluoro-alpha-methylbenzylamine, 4-fluoro-a-methylbenzylamine, 1-p-fluorophenyl-ethylamine, 1-4-fluorophenyl ethan-1-amine, p-fluoro-a-methylbenzylamine, rs-1-4-fluorophenyl ethylamine
123063
1-(4-fluorophenyl)ethanamine
CC(C1=CC=C(C=C1)F)N

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point -30°C
Color Colorless to Yellow or Orange
Density 1.0590g/mL
Boiling Point 97°C (35.0 mmHg)
Flash Point 68°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula FC6H4CH(CH3)NH2
Refractive Index 1.5005 to 1.5025
Quantity 1 g
Specific Gravity 1.059
Formula Weight 139.17
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material DL-4-Fluoro-α-methylbenzylamine, 97%
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'