missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-3-Methyl-2-butanol, 98%, Thermo Scientific Chemicals
10.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Colorless |
Density | 0.8100g/mL |
Boiling Point | 112.0°C |
Flash Point | 39°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98% |
Packaging | Glass bottle |
Linear Formula | (CH3)2CHCH(OH)CH3 |
Refractive Index | 1.4080 to 1.4100 |
Quantity | 25 mL |
Show More |
Safety and Handling
Product Identifier
- DL-3-Methyl-2-butanol
- Warning
- Acute toxicity Category 4
- Flammable liquid Category 3
- H332-Harmful if inhaled.
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'