missing translation for 'onlineSavingsMsg'
Learn More

DL-2-Phenylpropionic acid, 98%, ACROS Organics™

Brand:  Acros Organics 130871000

Code : 29

Additional Details : CAS Number : 492-37-5 Weight : 0.15000kg


Quantity 100g
Packaging Glass bottle

 View more versions of this product

Product Code. 10666734

  • £185.40 / 100g


Estimated Shipment
18-06-2018
Add to basket


Specifications

Chemical Identifiers

DL-2-Phenylpropionic acid
98%
CH3CH(C6H5)CO2H
MFCD00002650
≥97.5%
C9H10O2
150.18
DL-Hydratropic acid

Specifications

DL-2-Phenylpropionic acid
Clear pale yellow to yellow
492-37-5
Glass bottle
0.3% max.
≥97.5%
C9H10O2
150.18
09, 524
1.1g/mL
100g
Authentic
1.5215 to 1.5235 (20°C, 589nm)
Liquid
98%
CH3CH(C6H5)CO2H
MFCD00002650
DL-Hydratropic acid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • DL-2-Phenylpropionic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS