missing translation for 'onlineSavingsMsg'
Learn More

DL-2-Phenylpropionaldehyde, 98%, Thermo Scientific Chemicals

Catalog Number p-3779439
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
93-53-8
C9H10O
134.18
MFCD00006973
IQVAERDLDAZARL-UHFFFAOYSA-N
2-phenylpropionaldehyde, hydratropaldehyde, hydratropic aldehyde, hyacinthal, cumene aldehyde, hydratropa aldehyde, hydrotropic aldehyde, 2-phenyl-1-propanal, 2-phenyl propionaldehyde, aldehyd hydratropovy
7146
2-phenylpropanal
CC(C=O)C1=CC=CC=C1
This item is not returnable. View return policy
93-53-8
C9H10O
134.18
MFCD00006973
IQVAERDLDAZARL-UHFFFAOYSA-N
2-phenylpropionaldehyde, hydratropaldehyde, hydratropic aldehyde, hyacinthal, cumene aldehyde, hydratropa aldehyde, hydrotropic aldehyde, 2-phenyl-1-propanal, 2-phenyl propionaldehyde, aldehyd hydratropovy
7146
2-phenylpropanal
CC(C=O)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -60.0°C
Color Colorless
Density 1.0110g/mL
Boiling Point 202.0°C to 205.0°C
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH(C6H5)CHO
Refractive Index 1.5160 to 1.5240
Quantity 25 g
Beilstein 07, 305
Specific Gravity 1.011
Solubility Information Solubility in water: 3.4g/L (20°C).
Viscosity 2.16 mPa.s (20°C)
Formula Weight 134.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material DL-2-Phenylpropionaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • DL-2-Phenylpropionaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'