missing translation for 'onlineSavingsMsg'
Learn More

DL-2-Phenylglycine, 98%, Thermo Scientific™

Catalog Number 11444617
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
2835-06-5
C8H9NO2
151.17
MFCD00064402
This item is not returnable. View return policy
2835-06-5
C8H9NO2
151.17
MFCD00064402

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point ∼290°C (sublimation)
Beilstein 3197862
Merck Index 14,7291
Synonym 5-amino-2-methyl-benzoic acid, 5-amino-2-methyl benzoic acid, 2-methyl-5-aminobenzoic acid, 5-amino-o-toluic acid, 5-amino-2-methylbenzoicacid, benzoic acid, 5-amino-2-methyl, pubchem4980, 3-amino-6-methylbenzoic acid, ksc494q3t, 2-methyl-5-amino benzoic acid
InChI Key FSXVZWAWYKMFMX-UHFFFAOYSA-N
SMILES CC1=C(C=C(C=C1)N)C(=O)O
IUPAC Name 5-amino-2-methylbenzoic acid
PubChem CID 10374614
Formula Weight 151.17
Percent Purity 98%
Quantity 100 g
Odor Odorless
Chemical Name or Material DL-2-Phenylglycine
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'