missing translation for 'onlineSavingsMsg'
Learn More

DL-2-Methyl-1-butanol, 98%, Thermo Scientific Chemicals

Catalog Number p-160858
Click to view available options
Quantity:
25 mL
100 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
137-32-6
C5H12O
88.15
MFCD00004743
QPRQEDXDYOZYLA-UHFFFAOYSA-N
2-methyl-1-butanol, active amyl alcohol, 1-butanol, 2-methyl, 2-methylbutanol, sec-butylcarbinol, dl-2-methyl-1-butanol, 2-methyl-n-butanol, 2-methylbutyl alcohol, 2-methyl butanol-1, primary active amyl alcohol
8723
CHEBI:48945
2-methylbutan-1-ol
CCC(C)CO
This item is not returnable. View return policy
137-32-6
C5H12O
88.15
MFCD00004743
QPRQEDXDYOZYLA-UHFFFAOYSA-N
2-methyl-1-butanol, active amyl alcohol, 1-butanol, 2-methyl, 2-methylbutanol, sec-butylcarbinol, dl-2-methyl-1-butanol, 2-methyl-n-butanol, 2-methylbutyl alcohol, 2-methyl butanol-1, primary active amyl alcohol
8723
CHEBI:48945
2-methylbutan-1-ol
CCC(C)CO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -70.0°C
Color Colorless to Yellow
Density 0.8100g/mL
Boiling Point 130.0°C
Flash Point 43°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C2H5CH(CH3)CH2OH
Refractive Index 1.4090 to 1.4110
Quantity 25 mL
Beilstein 01, 388
Merck Index 15, 6104
Specific Gravity 0.81
Solubility Information Solubility in water: 3.6g/100mL (30°C). Other solubilities: miscible with alcohol and ether,soluble in most organic solvents
Viscosity 5.1 mPa.s (20°C)
Formula Weight 88.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material DL-2-Methyl-1-butanol, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'