DL-2-Aminoadipic acid hydrate, 97%, ACROS Organics™


Overview
£44.00 valid until 31/03/2017
Log in to get the promotional price



Brand: Acros Organics

Manufacturer Part Number: 154500050

5GR DL-2-Aminoadipic acid hydrate, 97%

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 542-32-5 Weight: 0.06000kg


Packaging Glass bottle
Quantity 5g

Product Code. 10113160

Quantity Price
1 £ 110.0 / 5g
Estimated Shipment date
from Supplier 06-03-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material DL-2-Aminoadipic acid hydrate
CAS 542-32-5
CAS na
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H11NO4·xH2O
Linear Formula HO2C(CH2)3CH(NH2)CO2H·xH2O
Formula Weight 161.16
MDL Number MFCD00150708

Specification

Physical Form Powder
Assay ≥96.0%
Formula Weight 161.16
Quantity 5g
Color White
Beilstein 04, 495
Merck Index 14, 416
Linear Formula HO2C(CH2)3CH(NH2)CO2H·xH2O
CAS 542-32-5
Infrared Spectrum Authentic
Assay Percent Range 97%
MDL Number MFCD00150708
Packaging Glass bottle
Molecular Formula C6H11NO4·xH2O
Chemical Name or Material DL-2-Aminoadipic acid hydrate
CAS na
Documentation

Technical documents