missing translation for 'onlineSavingsMsg'
Learn More

DL-2-Amino-3-methyl-1-butanol, 97%, Thermo Scientific Chemicals

Catalog Number p-57383
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
16369-05-4
MFCD00004730
NWYYWIJOWOLJNR-UHFFFAOYSA-N
2-amino-3-methyl-1-butanol, dl-valinol, valinol, l-+-valinol, 2-amino-3-methyl-butan-1-ol, 1-butanol, 2-amino-3-methyl, 1-butanol, 2-amino-3-methyl-, .+/-., s-+-2-amino-3-methyl-1-butanol, 1-2-amino-3-methylbutan-1-ol
79019
2-amino-3-methylbutan-1-ol
CC(C)C(CO)N
This item is not returnable. View return policy
16369-05-4
MFCD00004730
NWYYWIJOWOLJNR-UHFFFAOYSA-N
2-amino-3-methyl-1-butanol, dl-valinol, valinol, l-+-valinol, 2-amino-3-methyl-butan-1-ol, 1-butanol, 2-amino-3-methyl, 1-butanol, 2-amino-3-methyl-, .+/-., s-+-2-amino-3-methyl-1-butanol, 1-2-amino-3-methylbutan-1-ol
79019
2-amino-3-methylbutan-1-ol
CC(C)C(CO)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Molecular Formula C5H13NO
Linear Formula (CH3)2CHCH(NH2)CH2OH
Refractive Index 1.4533 to 1.4553 (20°C, 589nm)
Quantity 1 g
Molecular Weight (g/mol) 103.165
Formula Weight 103.16
Percent Purity ≥94%
Physical Form Liquid
Chemical Name or Material DL-2-Amino-3-methyl-1-butanol, 95%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'