missing translation for 'onlineSavingsMsg'
Learn More

DL-2-Amino-1-propanol, 98%, ACROS Organics™

Catalog Number p-58683
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
6168-72-5
C3H9NO
75.11
MFCD00008085
BKMMTJMQCTUHRP-UHFFFAOYNA-N
dl-alaninol, 2-aminopropanol, 2-amino-1-propanol, dl-2-amino-1-propanol, 1-propanol, 2-amino, 1-hydroxy-2-aminopropane, 2-amino-2-methylethanol, alaninol, propanol, 2-amino, 1-methyl-2-hydroxyethylamine
5126
2-aminopropan-1-ol
CC(N)CO
This item is not returnable. View return policy
6168-72-5
C3H9NO
75.11
MFCD00008085
BKMMTJMQCTUHRP-UHFFFAOYNA-N
dl-alaninol, 2-aminopropanol, 2-amino-1-propanol, dl-2-amino-1-propanol, 1-propanol, 2-amino, 1-hydroxy-2-aminopropane, 2-amino-2-methylethanol, alaninol, propanol, 2-amino, 1-methyl-2-hydroxyethylamine
5126
2-aminopropan-1-ol
CC(N)CO
Melting Point 8.0°C
Color Colorless to Yellow
Density 0.9430g/mL
Boiling Point 173.0°C to 176.0°C
Flash Point 83°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3CH(NH2)CH2OH
Refractive Index 1.4480 to 1.4510
Quantity 5 g
Beilstein 04, I, 432
Merck Index 10, 472
Specific Rotation -0.5° to +0.5° (20°C, 589nm) (neat)
Solubility Information Solubility in water: freely soluble. Other solubilities: freely soluble in alcohol and ether
Formula Weight 75.11
Percent Purity 98%
Physical Form Viscous Liquid
Chemical Name or Material DL-2-Amino-1-propanol, 98%
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'