missing translation for 'onlineSavingsMsg'
Learn More

DL-2,3-Diaminopropionic acid monohydrochloride, 99%, ACROS Organics™

£52.57 - £181.20

Chemical Identifiers

CAS 54897-59-5
Molecular Formula C3H8N2O2·HCl
Molecular Weight (g/mol) 140.57
MDL Number MFCD00012884
InChI Key SKWCZPYWFRTSDD-UHFFFAOYSA-N
Synonym 2,3-diaminopropanoic acid hydrochloride, dl-2,3-diaminopropionic acid hydrochloride, 3-amino-dl-alanine hydrochloride, dl-2,3-diaminopropionic acid monohydrochloride, 2,3-diaminopropionic acid hydrochloride, +--2,3-diaminopropionic acid hydrochloride, 3-amino-dl-alanine monohydrochloride, 2,3-diaminopropionic acid monohydrochloride, dl-2,3-diaminopropanoic acid hydrochloride, diaminopropionic acid
PubChem CID 108638
IUPAC Name 2,3-diaminopropanoic acid;hydrochloride
SMILES C(C(C(=O)O)N)N.Cl
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10062680
SDS View Documents
Acros Organics
204670050
5g Glass bottle £52.57
5g View alternative products
Save Up to 
»
Estimated Shipment
02-11-2020
Please to purchase this item. Need a web account? Register with us today!
10299403
SDS View Documents
Acros Organics
204670250
25g Glass bottle £181.20
25g View alternative products
Save Up to 
»
Estimated Shipment
02-11-2020
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

54897-59-5
140.57
SKWCZPYWFRTSDD-UHFFFAOYSA-N
108638
C(C(C(=O)O)N)N.Cl
C3H8N2O2·HCl
MFCD00012884
2,3-diaminopropanoic acid hydrochloride, dl-2,3-diaminopropionic acid hydrochloride, 3-amino-dl-alanine hydrochloride, dl-2,3-diaminopropionic acid monohydrochloride, 2,3-diaminopropionic acid hydrochloride, +--2,3-diaminopropionic acid hydrochloride, 3-amino-dl-alanine monohydrochloride, 2,3-diaminopropionic acid monohydrochloride, dl-2,3-diaminopropanoic acid hydrochloride, diaminopropionic acid
2,3-diaminopropanoic acid;hydrochloride

Specifications

DL-2, 3-Diaminopropionic acid monohydrochloride
Glass bottle
222°C to 226°C
54897-59-5
C3H8N2O2·HCl
MFCD00012884
10, 2948
Solubility in water: freely soluble in water
C(C(C(=O)O)N)N.Cl
140.57
140.57
99%
Authentic
White to Beige
5g
98.5% min. (Argentometry)
H2NCH2CH(NH2)CO2H·HCl
04, 405
2,3-diaminopropanoic acid hydrochloride, dl-2,3-diaminopropionic acid hydrochloride, 3-amino-dl-alanine hydrochloride, dl-2,3-diaminopropionic acid monohydrochloride, 2,3-diaminopropionic acid hydrochloride, +--2,3-diaminopropionic acid hydrochloride, 3-amino-dl-alanine monohydrochloride, 2,3-diaminopropionic acid monohydrochloride, dl-2,3-diaminopropanoic acid hydrochloride, diaminopropionic acid
SKWCZPYWFRTSDD-UHFFFAOYSA-N
2,3-diaminopropanoic acid;hydrochloride
108638
Fine Crystalline Powder
Safety and Handling

Safety and Handling

Product Identifier
  • DL-2,3-Diaminopropionic acid monohydrochloride

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.