missing translation for 'onlineSavingsMsg'
Learn More

DL-1-(2-Furyl)ethanol, 99%, stabilized, Thermo Scientific Chemicals

Catalog Number p-137350
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
4208-64-4
C6H8O2
112.13
MFCD00077775
UABXUIWIFUZYQK-UHFFFAOYSA-N
1-2-furyl ethanol, 1-furan-2-yl ethanol, 1-furan-2-yl-ethanol, 1-2-furyl ethan-1-ol, 2-furanmethanol, ?-methyl, 1-furan-2-yl ethan-1-ol, 2-furanmethanol,, a-methyl, methylfurfuryl alcohol, alpha-methylfuran-2-methanol, 1-furylethanol
107243
1-(furan-2-yl)ethanol
CC(C1=CC=CO1)O
This item is not returnable. View return policy
4208-64-4
C6H8O2
112.13
MFCD00077775
UABXUIWIFUZYQK-UHFFFAOYSA-N
1-2-furyl ethanol, 1-furan-2-yl ethanol, 1-furan-2-yl-ethanol, 1-2-furyl ethan-1-ol, 2-furanmethanol, ?-methyl, 1-furan-2-yl ethan-1-ol, 2-furanmethanol,, a-methyl, methylfurfuryl alcohol, alpha-methylfuran-2-methanol, 1-furylethanol
107243
1-(furan-2-yl)ethanol
CC(C1=CC=CO1)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.0700g/mL
Boiling Point 72°C to 74°C (15.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Refractive Index 1.4770 to 1.48
Quantity 1 g
Beilstein 17, V,3, 374
Specific Gravity 1.07
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in ethanol and most common organic solvent
Stabilizer 0.3 to 0.5% hydroquinone
Formula Weight 112.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material DL-1-(2-Furyl)ethanol, 99%
Show More Show Less
compliance-icons
Product Identifier
  • DL-1-(2-Furyl)ethanol, stabilized
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'