missing translation for 'onlineSavingsMsg'
Learn More

Divinyltetramethyldisiloxane, 97%, Thermo Scientific Chemicals

Catalog Number 10736592
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2627-95-4
C8H18OSi2
186.40
MFCD00014933
BITPLIXHRASDQB-UHFFFAOYSA-N
1,3-divinyltetramethyldisiloxane, 1,1,3,3-tetramethyl-1,3-divinyldisiloxane, disiloxane, 1,3-diethenyl-1,1,3,3-tetramethyl, 1,1'-divinyltetramethyldisiloxane, unii-sj4d71zqf4, bisvinyltetramethyldisiloxane, 1,3-divinyl-1,1,3,3-tetramethyl disiloxane, sym-tetramethyldivinyldisiloxane, tetramethyl-1,3-divinyldisiloxane, 1,3-diethenyl-1,1,3,3-tetramethyldisiloxane
75819
ethenyl-[ethenyl(dimethyl)silyl]oxy-dimethylsilane
C[Si](C)(O[Si](C)(C)C=C)C=C
This item is not returnable. View return policy
2627-95-4
C8H18OSi2
186.40
MFCD00014933
BITPLIXHRASDQB-UHFFFAOYSA-N
1,3-divinyltetramethyldisiloxane, 1,1,3,3-tetramethyl-1,3-divinyldisiloxane, disiloxane, 1,3-diethenyl-1,1,3,3-tetramethyl, 1,1'-divinyltetramethyldisiloxane, unii-sj4d71zqf4, bisvinyltetramethyldisiloxane, 1,3-divinyl-1,1,3,3-tetramethyl disiloxane, sym-tetramethyldivinyldisiloxane, tetramethyl-1,3-divinyldisiloxane, 1,3-diethenyl-1,1,3,3-tetramethyldisiloxane
75819
ethenyl-[ethenyl(dimethyl)silyl]oxy-dimethylsilane
C[Si](C)(O[Si](C)(C)C=C)C=C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -99.7°C
Color Colorless
Density 0.8130g/mL
Boiling Point 139°C
Flash Point 24°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula [H2C=CHSi(CH3)2]2O
Refractive Index 1.4110 to 1.413
Quantity 10 g
Specific Gravity 0.813
Solubility Information Solubility in water: insoluble
Formula Weight 186.4
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Divinyltetramethyldisiloxane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Divinyltetramethyldisiloxane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'