missing translation for 'onlineSavingsMsg'
Learn More

Divinylbenzene, 80%, mixture of isomers, stab. with 1000ppm 4-tert-butylcatechol, Thermo Scientific Chemicals

Catalog Number 11369977
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
1321-74-0
C10H10
130.19
MFCD01778656,MFCD00010654
MYRTYDVEIRVNKP-UHFFFAOYSA-N
divinylbenzene, 1,2-divinylbenzene, o-divinylbenzene, benzene, diethenyl, benzene, 1,2-diethenyl, divinylbenzen, divinyl benzen, divinyl-benzene, poly divinylbenzene, divinylbenzene dvb
66666
1,2-bis(ethenyl)benzene
C=CC1=CC=CC=C1C=C
This item is not returnable. View return policy
1321-74-0
C10H10
130.19
MFCD01778656,MFCD00010654
MYRTYDVEIRVNKP-UHFFFAOYSA-N
divinylbenzene, 1,2-divinylbenzene, o-divinylbenzene, benzene, diethenyl, benzene, 1,2-diethenyl, divinylbenzen, divinyl benzen, divinyl-benzene, poly divinylbenzene, divinylbenzene dvb
66666
1,2-bis(ethenyl)benzene
C=CC1=CC=CC=C1C=C

Divinylbenzene is used as a reactive monomer in polyester resins and for the synthesis of highly crosslinked macroporous styrene as well as a crosslinking agent. It acts as an intermediate in the production of ion exchange resin. Further, it is used in making rubber, petroleum production and polymeric core shell nanoparticles. In addition, it is used to synthesize monodispersed poly divinylbenzene microspheres.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 0.914 g/mL
Flash Point 69°C (156°F)
Refractive Index 1.574
Quantity 25 g
Sensitivity Light sensitive
Solubility Information Miscible with ethanol and ether. Immiscible with water.
Formula Weight 130.19
Percent Purity 80%
Physical Form Liquid
Chemical Name or Material Divinylbenzene, Mixture of isomers, Stabilized with 1000ppm 4-tert-butylcatechol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'