missing translation for 'onlineSavingsMsg'
Learn More

Dithiooxamide, 98%, Thermo Scientific Chemicals

Catalog Number p-7027831
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
79-40-3
C2H4N2S2
120.188
MFCD00004941
OAEGRYMCJYIXQT-UHFFFAOYSA-N
dithiooxamide, rubeanic acid, ethanebis thioamide, oxamide, dithio, dithioxamide, rubeane, rubean, hydrorubeanic acid, oxaldiimidic acid, dithio, usaf mk-6
2777982
ethanedithioamide
C(=S)(C(=S)N)N
This item is not returnable. View return policy
79-40-3
C2H4N2S2
120.188
MFCD00004941
OAEGRYMCJYIXQT-UHFFFAOYSA-N
dithiooxamide, rubeanic acid, ethanebis thioamide, oxamide, dithio, dithioxamide, rubeane, rubean, hydrorubeanic acid, oxaldiimidic acid, dithio, usaf mk-6
2777982
ethanedithioamide
C(=S)(C(=S)N)N

Dithiooxamide acts as a chelating agent and used in the determination of copper(II), nickel(II) and cobalt(II). It is used as a building block in the synthesis of cyclen. It is involved in the preparation of thiazolothiazole-linked porous organic polymers and N,N'-disubstituted dithiooxamides. It is also employed as a modifier to prepare the modified glassy carbon electrode which is used to investigate the electrochemical properties of quercetin, an important flavonoid derivative. Further, it is involved in the preparation of chelating resin with formaldehyde, which finds application in separation and concentration of silver ions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dithiooxamide acts as a chelating agent and used in the determination of copper(II), nickel(II) and cobalt(II). It is used as a building block in the synthesis of cyclen. It is involved in the preparation of thiazolothiazole-linked porous organic polymers and N,N′-disubstituted dithiooxamides. It is also employed as a modifier to prepare the modified glassy carbon electrode which is used to investigate the electrochemical properties of quercetin, an important flavonoid derivative. Further, it is involved in the preparation of chelating resin with formaldehyde, which finds application in separation and concentration of silver ions.

Solubility
Soluble in acetone and chloroform. Insoluble in water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 245°C (decomposition)
Quantity 5 g
Solubility Information Soluble in acetone and chloroform. Insoluble in water.
Formula Weight 120.2
Percent Purity 98%
Chemical Name or Material Dithiooxamide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'