missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palintest™ Photometer Tube Test
Predispensed reagent tests require minimal handling and speed up processing. Palintest™; Photometer Tube Test reagents are supplied in 16mm tubes for improved resolution, with greater sensitivity in the lower part of the measurement range.
£98.83 - £116.26
Specifications
For Use With (Equipment) | Photometer, heater block |
---|---|
Type | Photometer tube test reagent |
Product Code | Brand | No. of Tests | Range | Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Tests | Range | Type | Price | Quantity & Availability | |||
11988987
|
Palintest™
PL 503 |
30 | 0 to 2.0mg/L | Photometer tube test reagent |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
13191461
|
Palintest™
PL 513 |
30 | 0 to 20mg/L | Photometer tube test reagent |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11998987
|
Palintest™
PL 513 |
30 | 0 to 20mg/L | Photometer tube test reagent |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
13181461
|
Palintest™
PL 503 |
30 | 0 to 2.0mg/L | Photometer tube test reagent |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
- For ultimate convenience, the Palintest™ range of Tubetest reagents offer pre-dispensed reagents for minimal handling with barcoded reagent labelling for ease of use in wastewater applications.
- For a complete wastewater analysis system, use Palintest™ tubetests with any Palintest™ multiparameter photometer and heater block.
- Tubetest reagents are supplied in 16mm tubes for improved resolution, with greater sensitivity in the lower part of the measurement range.
Specifications
Photometer, heater block | |
Photometer tube test reagent |