Learn More
Fisherbrand™ Dissecting Scissors
Use these scissors for a variety of applications. These dissecting scissors are suitable for theatre, ward, veterinary, dental and chiropody use.
Brand:  Fisherbrand™
Code : NEW
Additional Details : Weight : 0.15480kg
 View more versions of this product
Product Code. 15277168
  • £49.05 / Pack of 10
In Stock
to see stock.
Add to basket
Description
Description
  • Stainless steel dissecting scissors
  • Closed shanks, fine point
  • Comply with the essential requirements of the Medical Devices Directive 93/42/EEC
  • Full traceability maintained throughout the manufacturing process.
  • Made from only high grade stainless steel in accordance to the specifications of ISO 7153-1
  • 10 year guarantee
Specifications
Specifications
Dissecting Scissors
Curved
105 mm
Stainless Steel