Learn More
Copan Italia™ Disposable Pasteur Pipets
Use Copan Italia™ Disposable Pasteur Pipets for biological fluids and most acids. Their low surface affinity reduces the loss of cells and proteins of interest.
POA
Specifications
Bulb Draw (Metric) | 3.5 mL |
---|---|
Material | LDPE |
For Use With (Application) | For transferring and dispensing liquids safely in all types of laboratories |
Graduated | Yes |
Type | Pasteur Pipette |
Description
- Unbreakable
- Can be sealed and frozen
- Usable in liquid nitrogen
- Sterile
- Polyethylene LDPE
Specifications
3.5 mL | |
For transferring and dispensing liquids safely in all types of laboratories | |
Pasteur Pipette |
LDPE | |
Yes | |
Single-use |