missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Disposable IR Cards, 5x10cm, NaCl window, 9.5mm aperture

Provide a conveniently mounted, non-absorbing substrate for qualitative infrared analysis.

Brand:  Alfa Aesar™ 043931.KM

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15456565

  • £155.60 / Pack of 25


Estimated Shipment date
from Supplier 02-07-2018
Add to basket

Description

Description

  • Size: 5 x 10cm
  • NaCl window
  • 9.5mm aperture
Specifications

Specifications

Disposable IR Cards
25 Pk.
Videos
SDS