Learn More
Alfa Aesar™ Disposable IR Cards, 5x10cm, KCl window, 15mm aperture
Provide a conveniently mounted, non-absorbing substrate for qualitative infrared analysis.
Brand:  Alfa Aesar™ 043933.KJ
Code : NEW
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15486565
  • £95.19 / Pack of 5
Estimated Shipment date
from Supplier 05-02-2018
Add to basket
Specifications
Specifications
Disposable IR Cards
5 Pk.