Learn More
Fisherbrand™ Disposable Inoculating Loops and Needles
Designed for quantitative procedures such as sampling, serial dilutions, as well as for bacterial inoculation. Fisherbrand™ Disposable Inoculating Loops and Needles, Length: 8 in. (20cm) are precision molded from a specially formulated, biologically inert polymer.
Brand: Fisherbrand™
Code : NEW
Additional Details : Weight : 7.00000kg
Description
Designed for quantitative procedures such as sampling, serial dilutions, as well as for bacterial inoculation. Fisherbrand™ Disposable Inoculating Loops and Needles, Length: 8 in. (20cm) are precision molded from a specially formulated, biologically inert polymer.
Ideal for anaerobic microbiology applications where deep agar inoculation is required. Use in situations where flaming is not practical, such as under hoods and in anaerobic chambers. Flexible enough to facilitate streaking, yet rigid enough to penetrate the media for subsurface inoculation.- Combi Loop™ has a 10μL loop on one end and a 1μL loop on the other
- Convenient than metal loops; use a loop directly from the package, then throw it away
- Gamma irradiated for sterility in accordance with U.S. Pharmacopeia Volume XX
- No flaming required, eliminating the hazards of open flames, spattering, cross-contamination, and the risk of infection from pathogens distributed by aerosol formation
- Color coded for easy identification
Specifications
Yes | |
Biologically Inert Polymer | |
Sterile | |
20 cm | |
Natural | |
Certificate of dimensional accuracy | |
Ziplock Bag |
0.09/0.38 cm | |
8 in. | |
0.035/0.14 in. | |
0.1 μL | |
Inoculating Loop and Needle, Flexible | |
5000/Cs. |