missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Tilt Measure Dispenser with Bottle

Particularly well-suited for dispensing suspensions. Comes complete with 1000mL soda-lime glass reservoir bottle.

Brand:  BRAND™ 43005

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10128371

  • £60.46 / Each


Estimated Shipment: 15-11-2021
Add to basket

Description

Description

  • Dispenser head made from Borosilicate glass (Boro 3.3)
  • Calibrated to deliver (TD, Ex)
  • Inscriptions in green enamel
  • Particularly well-suited for dispensing suspensions
  • Complete with 1000mL soda-lime glass reservoir bottle with 29/32 neck size
Specifications

Specifications

Clear
5 mL
1000mL soda-lime glass reserve bottle
Top Tilt Pipette Dispenser
Borosilicate Glass
Videos
SDS
Documents
Certificates