missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Tilt Measure Dispenser with Bottle

Particularly well-suited for dispensing suspensions. Comes complete with 1000mL soda-lime glass reservoir bottle.

Brand:  BRAND™ 43008

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10138371

Quantity Price
1 £84.65 / Each
10 £68.20 / Each
Estimated Shipment: 17-11-2021

Add to basket


Description

Description

  • Dispenser head made from Borosilicate glass (Boro 3.3)
  • Calibrated to deliver (TD, Ex)
  • Inscriptions in green enamel
  • Particularly well-suited for dispensing suspensions
  • Complete with 1000mL soda-lime glass reservoir bottle with 29/32 neck size
Specifications

Specifications

Clear
10 mL
1000mL soda-lime glass reserve bottle
Top Tilt Pipette Dispenser
Borosilicate Glass
Videos
SDS
Documents
Certificates