Learn More
Brand™ Dispensette™ S Organic Analog-adjustable Bottletop Dispensers, Exchange Program
Features a 'floating piston' design for low-force operation and no wearing parts. Optimized flow channels make dispensing and priming even easier-particularly in instruments with large volumes. Designed for dispensing of organic solvents, concentrated acids and peroxides.
POA
Specifications
Type Dispenser
Autoclavable Autoclavable at 121°C
Color Yellow
Display Analog
Description With recirculation valve
View More Specs
Description
Description
  • New discharge tube, with or without recirculation valve
  • New valve system - no additional sealing rings required, facilitates cleaning and preparations for autoclaving
  • Simple priming by optimized flow channels
  • Less dispensing force - especially with large volume instruments
  • Safe volume setting with internal scalloped track (analog-adjustable instruments)
  • Pivoted screw cap - does not disturb dispensing
  • Easy to calibrate and adjust in order to comply with GLP guidelines
  • Easy to dismantle for cleaning
  • Replacable filling and discharge valve with safety bulb (locks when discharge tube is not assembled)
  • 360° rotatable valve block allows bottle label to face user
  • Telescoping filling tube adjusts easily to different size bottles
  • DE-M marking
  • Exchange promotion
Specifications
Specifications
Dispenser
Yellow
With recirculation valve
Autoclavable at 121°C
Analog
ISO 9001